missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 5446-02-6 |
|---|---|
| Molecular Formula | C9H10O4 |
| Molecular Weight (g/mol) | 182.18 |
| MDL Number | MFCD00008424 |
| InChI Key | ZICRWXFGZCVTBZ-UHFFFAOYSA-N |
| Synonym | Methyle2-hydroxy-4-methoxybenzoate |
| IUPAC Name | methyl 2-hydroxy-4-methoxybenzoate |
| SMILES | COC(=O)C1=CC=C(OC)C=C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC174270500
|
N/A | 50g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 5446-02-6 | |
| 182.18 | |
| ZICRWXFGZCVTBZ-UHFFFAOYSA-N | |
| methyl 2-hydroxy-4-methoxybenzoate |
| C9H10O4 | |
| MFCD00008424 | |
| Methyle2-hydroxy-4-methoxybenzoate | |
| COC(=O)C1=CC=C(OC)C=C1O |
Specifications
| 5446-02-6 | |
| Authentic | |
| C9H10O4 | |
| MFCD00008424 | |
| Methyle2-hydroxy-4-methoxybenzoate | |
| COC(=O)C1=CC=C(OC)C=C1O | |
| 182.18 | |
| 98% |
| >112°C | |
| 97.5% min. (GC) | |
| CH3OC6H3(OH)CO2CH3 | |
| 10, 381 | |
| ZICRWXFGZCVTBZ-UHFFFAOYSA-N | |
| methyl 2-hydroxy-4-methoxybenzoate | |
| 182.18 | |
| Methyl 4-methoxysalicylate |
Safety and Handling
EINECSNumber : 226-657-5
RUO – Research Use Only