missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxypyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. M25081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25081G 1 g
M25085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25081G Supplier TCI America Supplier No. M25081G

Chemical Identifiers

CAS 29681-43-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00956223
InChI Key OJDKENGKKYVJLY-UHFFFAOYSA-N
Synonym methyl 4-methoxypicolinate, 4-methoxy-pyridine-2-carboxylic acid methyl ester, 4-methoxy-pyridine-2-carboxylicacidmethylester, 2-pyridinecarboxylic acid, 4-methoxy-, methyl ester, 4-methoxypicolinic acid methyl ester, 4-methoxypyridine-2-carboxylic acid methyl ester, methyl 4-methoxy-2-pyridinecarboxylate, 4-methoxy-pyridine-2-carboxylic acid metyl ester, pubchem11083, acmc-1b7sb
PubChem CID 1519345
IUPAC Name methyl 4-methoxypyridine-2-carboxylate
SMILES COC1=CC(=NC=C1)C(=O)OC

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Methoxypyridine-2-carboxylate