missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientific™

Catalog No. AC325710010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325710010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC325710010 Supplier Thermo Scientific Chemicals Supplier No. 325710010

Chemical Identifiers

CAS 22027-52-7
Molecular Formula C13H14O5
Molecular Weight (g/mol) 250.25
MDL Number MFCD00800637
InChI Key GFDCYWYWNSTMDX-UHFFFAOYSA-N
PubChem CID 3864207
IUPAC Name methyl 4-(3-methoxy-3-oxopropanoyl)benzoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 81°C to 86°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Quantity 1 g
Synonym methyl 4-3-methoxy-3-oxopropanoyl benzoate, methyl 4-methoxycarbonylbenzoylacetate, 4-2-methoxycarbonyl-acetyl-benzoic acid methyl ester, 4-methoxycarbonyl-beta-oxobenzenepropanoic acid methyl ester, benzenepropanoic acid,4-methoxycarbonyl-a-oxo-, methyl ester, acmc-20efie, methyl 4-methoxycarbonyl benzoyl acetate, methyl4-methoxycarbonylbenzoylacetate, 4-methoxycarbonylbenzoyl acetic acid methyl ester, methyl 3-4-methoxycarbonyl phenyl-3-oxopropanoate
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in acetone,methanol
Formula Weight 236.22
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Methyl 4-methoxycarbonylbenzoylacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only