missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Isopropoxybenzoate 97.0+%, TCI America™

Catalog No. I05875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05875G Supplier TCI America Supplier No. I05875G
Only null left

Chemical Identifiers

CAS 35826-59-6
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00084855
InChI Key ROEMZKPINYTSKV-UHFFFAOYSA-N
Synonym 4-Isopropoxybenzoic Acid Methyl Ester
PubChem CID 3015807
IUPAC Name methyl 4-propan-2-yloxybenzoate
SMILES CC(C)OC1=CC=C(C=C1)C(=O)OC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Isopropoxybenzoate