missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobutyrate, 95%, stabilized, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 426370050

Catalog No. AC426370050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl 4-iodobutyrate
14273-85-9
80°C to 83°C (11.0mmHg)
Authentic
Glass bottle
I(CH2)3COOCH3
5g
methyl 4-iodobutyrate, butanoic acid, 4-iodo-, methyl ester, butyric acid, 4-iodo-, methyl ester, methyl-4-iodobutyrate, acmc-1bwlv, methyl gamma-iodobutyrate, 4-iodobutyric acid methyl ester, butanoic acid, 4-iodo-,methyl ester
COC(=O)CCCI
228.02
547991
95%
Stabilized
1.6890g/mL
99°C
94% min. (GC)
C5H9IO2
MFCD00674066
1.689
NBCIIVXSBPDKOM-UHFFFAOYSA-N
methyl 4-iodobutanoate
Copper
228.02
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only