missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-imidazolecarboxylate, 98%

Catalog No. AC427630010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427630010 1 g Glass bottle
AC427630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427630010 Supplier Thermo Scientific Chemicals Supplier No. 427630010

CAS: 17325-26-7 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17325-26-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00216584
InChI Key DVLGIQNHKLWSRU-UHFFFAOYSA-N
Synonym methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
PubChem CID 565661
IUPAC Name methyl 1H-imidazole-5-carboxylate
SMILES COC(=O)C1=CN=CN1

Specifications

Melting Point 154°C to 156°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 126.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-imidazolecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only