missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbenzoate, 98+%

Catalog No. p-7027383
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2409106 5 g
AAB2409114 25 g
AAB2409122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2409106 Supplier Thermo Scientific Chemicals Supplier No. B2409106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1571-08-0 | C9H8O3 | 164.16 g/mol

Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1571-08-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00006950
InChI Key FEIOASZZURHTHB-UHFFFAOYSA-N
Synonym methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
PubChem CID 15294
IUPAC Name methyl 4-formylbenzoate
SMILES COC(=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 59°C to 63°C
Density 1.2
Boiling Point 264°C to 266°C
Flash Point 128°C (262°F)
Quantity 5 g
Beilstein 775895
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material Methyl 4-formylbenzoate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-385-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only