missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formyl-1,2,5-trimethyl-1H-pyrrole-3-carboxylate, 95%, Thermo Scientific™

Catalog No. p-2705371
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC22504CB 250mg Amber Glass Bottle
CC22504DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC22504CB Supplier Thermo Scientific Chemicals Supplier No. CC22504CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175276-49-0
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00203864
InChI Key NEDLMVCFWFSFNJ-UHFFFAOYSA-N
Synonym methyl 4-formyl-1,2,5-trimethyl-1h-pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylicacid, 4-formyl-1,2,5-trimethyl-, methyl ester, 1h-pyrrole-3-carboxylicacid,4-formyl-1,2,5-trimethyl-,methyl ester
PubChem CID 2776538
IUPAC Name methyl 4-formyl-1,2,5-trimethylpyrrole-3-carboxylate
SMILES CC1=C(C(=C(N1C)C)C(=O)OC)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 195.22
Percent Purity 95%
Chemical Name or Material Methyl 4-formyl-1, 2, 5-trimethyl-1H-pyrrole-3-carboxylate
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.