missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (4-Fluorobenzoyl)acetate 98.0+%, TCI America™
$123.03 - $367.22
Chemical Identifiers
| CAS | 63131-29-3 |
|---|---|
| Molecular Formula | C10H9FO3 |
| Molecular Weight (g/mol) | 196.177 |
| MDL Number | MFCD00000355 |
| InChI Key | HGLVYXXPRSNKQN-UHFFFAOYSA-N |
| Synonym | methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6 |
| PubChem CID | 579425 |
| IUPAC Name | methyl 3-(4-fluorophenyl)-3-oxopropanoate |
| SMILES | COC(=O)CC(=O)C1=CC=C(C=C1)F |
Chemical Identifiers
| 63131-29-3 | |
| 196.177 | |
| HGLVYXXPRSNKQN-UHFFFAOYSA-N | |
| 579425 | |
| COC(=O)CC(=O)C1=CC=C(C=C1)F |
| C10H9FO3 | |
| MFCD00000355 | |
| methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6 | |
| methyl 3-(4-fluorophenyl)-3-oxopropanoate |
Specifications
| 63131-29-3 | |
| 91°C | |
| MFCD00000355 | |
| methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6 | |
| COC(=O)CC(=O)C1=CC=C(C=C1)F | |
| 196.177 | |
| 196.18 | |
| Liquid |
| Yellow | |
| C10H9FO3 | |
| 1 g | |
| HGLVYXXPRSNKQN-UHFFFAOYSA-N | |
| methyl 3-(4-fluorophenyl)-3-oxopropanoate | |
| 579425 | |
| ≥98.0% (GC) | |
| Methyl (4-Fluorobenzoyl)acetate |
Safety and Handling
TSCA : No