missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Ethynylbenzoate 98.0+%, TCI America™

Catalog No. M27681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27681G 1 g
M27685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27681G Supplier TCI America Supplier No. M27681G

Chemical Identifiers

CAS 3034-86-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00168820
InChI Key JPGRSTBIEYGVNO-UHFFFAOYSA-N
Synonym 4-ethynylbenzoic acid methyl ester, 4-ethynyl-benzoic acid methyl ester, benzoic acid, 4-ethynyl-, methyl ester, methyl4-ethynylbenzoate, 4-ethynylbenzoic acid methylester, 4-methoxycarbonyl phenylacetylene, benzoic acid,4-ethynyl-, methyl ester, methyl 4-ethynyl-benzoate, 4-ethinylbenzoic acid methyl ester, 4-ethynyl benzoic acid methylester
PubChem CID 640163
IUPAC Name methyl 4-ethynylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C#C

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Ethynylbenzoate
TSCA No
Recommended Storage Refrigerator