missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-cyano-5-(methylthio)thiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7028936
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2635303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2635303 Supplier Thermo Scientific Chemicals Supplier No. H2635303
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175202-48-9
Molecular Formula C8H7NO2S2
Molecular Weight (g/mol) 213.27
MDL Number MFCD00173757
InChI Key MJOQLNYXSSXZDJ-UHFFFAOYSA-N
Synonym methyl 4-cyano-5-methylthio thiophene-2-carboxylate, methyl 4-cyano-5-methylsulfanyl thiophene-2-carboxylate, 4-cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester, methyl 4-cyano-5-methylthiothiophene-2-carboxylate, 2-thiophenecarboxylicacid, 4-cyano-5-methylthio-, methyl ester, acmc-20akgh, methyl 3-cyano-2-methylthio thiophene-5-carboxylate, methyl 4-cyano-5-methylsulfanyl-2-thiophenecarboxylate, 4-cyano-5-methylthio thiophene-2-carboxylic acid methyl ester
PubChem CID 18536976
IUPAC Name methyl 4-cyano-5-methylsulfanylthiophene-2-carboxylate
SMILES COC(=O)C1=CC(C#N)=C(SC)S1

Specifications

Melting Point 117°C to 119°C
Quantity 1 g
UN Number UN3439
Formula Weight 213.27
Percent Purity 97%
Chemical Name or Material Methyl 4-cyano-5-(methylthio)thiophene-2-carboxylate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only