missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chlorocinnamate 98.0+%, TCI America™

Catalog No. C16725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16725G Supplier TCI America Supplier No. C16725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7560-44-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00075790
InChI Key IIBXQGYKZKOORG-QPJJXVBHSA-N
Synonym methyl 4-chlorocinnamate, methyl p-chlorocinnamate, methyl 3-4-chlorophenyl acrylate, 4-chlorocinnamic acid methyl ester, cinnamic acid, p-chloro-, methyl ester, 2-propenoic acid, 3-4-chlorophenyl-, methyl ester, methyl e-3-4-chlorophenyl prop-2-enoate, e-methyl 3-4-chlorophenyl acrylate, methyl 2e-3-4-chlorophenyl prop-2-enoate, methyl 3-4-chlorophenyl prop-2-enoate
PubChem CID 5314314
IUPAC Name methyl (E)-3-(4-chlorophenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(C=C1)Cl

Specifications

Melting Point 75°C
Color White
Quantity 5 g
Formula Weight 196.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Chlorocinnamate