missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorocinnamate, 99%

Catalog No. p-7023477
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1707106 5 g
AAA1707114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1707106 Supplier Thermo Scientific Chemicals Supplier No. A1707106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7560-44-3 | C10H9ClO2 | 196.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7560-44-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00075790
InChI Key IIBXQGYKZKOORG-QPJJXVBHSA-N
Synonym methyl 4-chlorocinnamate, methyl p-chlorocinnamate, methyl 3-4-chlorophenyl acrylate, 4-chlorocinnamic acid methyl ester, cinnamic acid, p-chloro-, methyl ester, 2-propenoic acid, 3-4-chlorophenyl-, methyl ester, methyl e-3-4-chlorophenyl prop-2-enoate, e-methyl 3-4-chlorophenyl acrylate, methyl 2e-3-4-chlorophenyl prop-2-enoate, methyl 3-4-chlorophenyl prop-2-enoate
PubChem CID 5314314
IUPAC Name methyl (E)-3-(4-chlorophenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(C=C1)Cl

Specifications

Melting Point 74°C to 77°C
Quantity 5 g
Beilstein 2045629
Formula Weight 196.64
Percent Purity 99%
Chemical Name or Material Methyl 4-chlorocinnamate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-451-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only