missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloroacetoacetate, 97%

CAS: 32807-28-6 | C5H7ClO3 | 150.56 g/mol

$142.90 - $142.90

Chemical Identifiers

CAS 32807-28-6
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000938
InChI Key HFLMYYLFSNEOOT-UHFFFAOYSA-N
Synonym methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
PubChem CID 36240
IUPAC Name methyl 4-chloro-3-oxobutanoate
SMILES COC(=O)CC(=O)CCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC241620250
SDS View Documents
Thermo Scientific Chemicals
241620250
25 mL
Each for $142.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32807-28-6
150.56
HFLMYYLFSNEOOT-UHFFFAOYSA-N
36240
COC(=O)CC(=O)CCl
C5H7ClO3
MFCD00000938
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
methyl 4-chloro-3-oxobutanoate

Specifications

32807-28-6
Orange-Yellow to Yellow
212.0°C to 213.0°C
Authentic
Glass bottle
1.4560 to 1.4580
MFCD00000938
1.28
Solubility in water: 71g/L (20 c)
COC(=O)CC(=O)CCl
150.56
150.56
Liquid
14.0°C to 16.0°C
1.2800g/mL
107°C
96% min. (GC)
C5H7ClO3
ClCH2C(O)CH2COOCH3
25 mL
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
HFLMYYLFSNEOOT-UHFFFAOYSA-N
methyl 4-chloro-3-oxobutanoate
36240
97%
Methyl 4-chloroacetoacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 251-230-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only