missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloroacetoacetate, 97%

Catalog No. AC241620250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC241620250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC241620250 Supplier Thermo Scientific Chemicals Supplier No. 241620250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32807-28-6 | C5H7ClO3 | 150.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32807-28-6
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000938
InChI Key HFLMYYLFSNEOOT-UHFFFAOYSA-N
Synonym methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
PubChem CID 36240
IUPAC Name methyl 4-chloro-3-oxobutanoate
SMILES COC(=O)CC(=O)CCl

Specifications

Melting Point 14.0°C to 16.0°C
Color Orange-Yellow to Yellow
Density 1.2800g/mL
Boiling Point 212.0°C to 213.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.4580
Linear Formula ClCH2C(O)CH2COOCH3
Quantity 25 mL
Specific Gravity 1.28
Solubility Information Solubility in water: 71g/L (20 c)
Formula Weight 150.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 4-chloroacetoacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 251-230-5

RUO – Research Use Only