missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloroacetoacetate, 97+%

Catalog No. AAH6470922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6470922 100 g
AAH6470914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6470922 Supplier Thermo Scientific Chemicals Supplier No. H6470922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32807-28-6 | C5H7ClO3 | 150.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32807-28-6
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000938
InChI Key HFLMYYLFSNEOOT-UHFFFAOYSA-N
Synonym methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
PubChem CID 36240
IUPAC Name methyl 4-chloro-3-oxobutanoate
SMILES COC(=O)CC(=O)CCl

Specifications

Color Colorless
Density 1.305 g/mL
Boiling Point 85°C (4 mmHg)
Flash Point 102°C (216°F)
Refractive Index 1.456
Quantity 100 g
UN Number UN2922
Beilstein 1759939
Formula Weight 150.56
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Methyl 4-chloroacetoacetate
Show More Show Less
Hazard Category H301-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 251-230-5
RTECSNumber AK5115000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only