missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Chloro-2-pyridinecarboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 59°C |
| Quantity | 5 g |
| Formula Weight | 171.58 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 4-Chloro-2-pyridinecarboxylate |
Safety and Handling
| TSCA | No |