missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chloro-2-pyridinecarboxylate 98.0+%, TCI America™

Catalog No. M18321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18321G 1 g
M18325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18321G Supplier TCI America Supplier No. M18321G
Only null left

Chemical Identifiers

CAS 24484-93-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD04116183
InChI Key VTENWIPSWAMPKI-UHFFFAOYSA-N
Synonym methyl 4-chloropicolinate, 4-chloro-pyridine-2-carboxylic acid methyl ester, 4-chloropicolinic acid methyl ester, methyl 4-chloro-2-pyridinecarboxylate, 4-chloropyridine-2-carboxylic acid methyl ester, 4-chloro-2-pyridinecarboxylic acid methyl ester, 2-pyridinecarboxylic acid, 4-chloro-, methyl ester, methyl-4-chlorpyridin-2-carboxylat, pubchem9577, acmc-209gcn
PubChem CID 820890
IUPAC Name methyl 4-chloropyridine-2-carboxylate
SMILES COC(=O)C1=NC=CC(=C1)Cl

Specifications

Melting Point 59°C
Quantity 1 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Chloro-2-pyridinecarboxylate