missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Chloro-2-methylbutyrate 98.0+%, TCI America™
$102.52 - $309.43
Chemical Identifiers
| CAS | 13888-03-4 |
|---|---|
| Molecular Formula | C6H11ClO2 |
| Molecular Weight (g/mol) | 150.602 |
| MDL Number | MFCD00198001 |
| InChI Key | ZDDYJHVTUSUPFC-UHFFFAOYSA-N |
| Synonym | 4-Chloro-2-methylbutyric Acid Methyl Ester |
| PubChem CID | 11263584 |
| IUPAC Name | methyl 4-chloro-2-methylbutanoate |
| SMILES | CC(CCCl)C(=O)OC |
Chemical Identifiers
| 13888-03-4 | |
| 150.602 | |
| ZDDYJHVTUSUPFC-UHFFFAOYSA-N | |
| 11263584 | |
| CC(CCCl)C(=O)OC |
| C6H11ClO2 | |
| MFCD00198001 | |
| 4-Chloro-2-methylbutyric Acid Methyl Ester | |
| methyl 4-chloro-2-methylbutanoate |
Specifications
| 13888-03-4 | |
| 81°C | |
| MFCD00198001 | |
| 4-Chloro-2-methylbutyric Acid Methyl Ester | |
| CC(CCCl)C(=O)OC | |
| 150.602 | |
| 150.60 | |
| Liquid |
| Colorless | |
| C6H11ClO2 | |
| 5 g | |
| ZDDYJHVTUSUPFC-UHFFFAOYSA-N | |
| methyl 4-chloro-2-methylbutanoate | |
| 11263584 | |
| ≥98.0% (GC) | |
| Methyl 4-Chloro-2-methylbutyrate |
Safety and Handling
TSCA : No