missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-3-hydroxythiophene-2-carboxylate, 97%

CAS: 95201-93-7 | C6H5BrO3S | 237.067 g/mol

$158.71 - $487.22

Chemical Identifiers

CAS 95201-93-7
Molecular Formula C6H5BrO3S
Molecular Weight (g/mol) 237.067
MDL Number MFCD00052081
InChI Key UFTXASQYKJFRKI-UHFFFAOYSA-N
Synonym 2z-4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylic acid, 4-bromo-3-hydroxy-, methyl ester, acmc-20akgk, maybridge1_004012, methyl 4-bromo-3-hydroxy-2-thiophenecarboxylate, methyl-3-hydroxy-4-bromo-2-thiophenecarboxylate, methyl-4-bromo-3-hydroxy-2-thiophenecarboxylate, 4-bromo-3-oxo-2-thienylidene-methoxy-methanolate, 4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylicacid,4-bromo-3-hydroxy-,methyl ester
PubChem CID 2777611
IUPAC Name methyl 4-bromo-3-hydroxythiophene-2-carboxylate
SMILES COC(=O)C1=C(C(=CS1)Br)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2024988
View Documents
Thermo Scientific Chemicals
B2024988
2.5 g
Each for $158.71
Only null left
 
AAB2024909
View Documents
Thermo Scientific Chemicals
B2024909
10 g
Each for $487.22
Only null left
 
Description

Description

The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

Solubility
Sparingly Soluble in water 0.95 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
Specifications

Chemical Identifiers

95201-93-7
237.067
UFTXASQYKJFRKI-UHFFFAOYSA-N
2777611
COC(=O)C1=C(C(=CS1)Br)O
C6H5BrO3S
MFCD00052081
2z-4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylic acid, 4-bromo-3-hydroxy-, methyl ester, acmc-20akgk, maybridge1_004012, methyl 4-bromo-3-hydroxy-2-thiophenecarboxylate, methyl-3-hydroxy-4-bromo-2-thiophenecarboxylate, methyl-4-bromo-3-hydroxy-2-thiophenecarboxylate, 4-bromo-3-oxo-2-thienylidene-methoxy-methanolate, 4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylicacid,4-bromo-3-hydroxy-,methyl ester
methyl 4-bromo-3-hydroxythiophene-2-carboxylate

Specifications

95201-93-7
C6H5BrO3S
2.5 g
Sparingly Soluble in water 0.95g/L @ 25°C.
COC(=O)C1=C(C(=CS1)Br)O
237.067
237.08
Methyl 4-bromo-3-hydroxythiophene-2-carboxylate
79°C to 80°C
MFCD00052081
2z-4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylic acid, 4-bromo-3-hydroxy-, methyl ester, acmc-20akgk, maybridge1_004012, methyl 4-bromo-3-hydroxy-2-thiophenecarboxylate, methyl-3-hydroxy-4-bromo-2-thiophenecarboxylate, methyl-4-bromo-3-hydroxy-2-thiophenecarboxylate, 4-bromo-3-oxo-2-thienylidene-methoxy-methanolate, 4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylicacid,4-bromo-3-hydroxy-,methyl ester
UFTXASQYKJFRKI-UHFFFAOYSA-N
methyl 4-bromo-3-hydroxythiophene-2-carboxylate
2777611
97%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only