missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-3-hydroxythiophene-2-carboxylate, 97%

Catalog No. AAB2024988
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2024988 2.5 g
AAB2024909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2024988 Supplier Thermo Scientific Chemicals Supplier No. B2024988
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95201-93-7 | C6H5BrO3S | 237.067 g/mol

The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

Solubility
Sparingly Soluble in water 0.95 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.

Chemical Identifiers

CAS 95201-93-7
Molecular Formula C6H5BrO3S
Molecular Weight (g/mol) 237.067
MDL Number MFCD00052081
InChI Key UFTXASQYKJFRKI-UHFFFAOYSA-N
Synonym 2z-4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylic acid, 4-bromo-3-hydroxy-, methyl ester, acmc-20akgk, maybridge1_004012, methyl 4-bromo-3-hydroxy-2-thiophenecarboxylate, methyl-3-hydroxy-4-bromo-2-thiophenecarboxylate, methyl-4-bromo-3-hydroxy-2-thiophenecarboxylate, 4-bromo-3-oxo-2-thienylidene-methoxy-methanolate, 4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylicacid,4-bromo-3-hydroxy-,methyl ester
PubChem CID 2777611
IUPAC Name methyl 4-bromo-3-hydroxythiophene-2-carboxylate
SMILES COC(=O)C1=C(C(=CS1)Br)O

Specifications

Melting Point 79°C to 80°C
Quantity 2.5 g
Solubility Information Sparingly Soluble in water 0.95g/L @ 25°C.
Formula Weight 237.08
Percent Purity 97%
Chemical Name or Material Methyl 4-bromo-3-hydroxythiophene-2-carboxylate
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only