missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Benzyloxybenzoate 98.0+%, TCI America™

Catalog No. M21805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21805G 5 g
M218025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21805G Supplier TCI America Supplier No. M21805G
Only null left

Chemical Identifiers

CAS 32122-11-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00017613
InChI Key ZLLQTDQOTFCCDF-UHFFFAOYSA-N
PubChem CID 299810
IUPAC Name methyl 4-(benzyloxy)benzoate
SMILES COC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 99°C
Color White
Quantity 5 g
Synonym methyl 4-benzyloxybenzoate, methyl 4-benzyloxy benzoate, 4-benzyloxybenzoic acid methyl ester, methyl4-benzyloxybenzoate, methyl 4-benzyloxy benzenecarboxylate, methyl 4-phenylmethoxy benzoate, methyl-4-benzyloxybenzoate, acmc-209hr7, 4-benzyloxy-benzoic acid methyl ester, benzoic acid,4-phenylmethoxy-, methyl ester
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Benzyloxybenzoate