missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Aminonicotinate 98.0+%, TCI America™
$279.60 - $1,114.70
Chemical Identifiers
| CAS | 16135-36-7 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00955996 |
| InChI Key | OCARFFAPQGYGBP-UHFFFAOYSA-N |
| Synonym | methyl 4-aminonicotinate, 4-aminonicotinic acid methyl ester, 4-amino-nicotinic acid methyl ester, 4-aminopyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylic acid, 4-amino-, methyl ester, nicotinic acid, 4-amino-, methyl ester, 4-aminonicotinicacidmethylester, methyl-4-aminonicotinate, ksc493a3l, tpc-py097 |
| PubChem CID | 592353 |
| IUPAC Name | methyl 4-aminopyridine-3-carboxylate |
| SMILES | COC(=O)C1=C(N)C=CN=C1 |
Chemical Identifiers
| 16135-36-7 | |
| 152.15 | |
| OCARFFAPQGYGBP-UHFFFAOYSA-N | |
| 592353 | |
| COC(=O)C1=C(N)C=CN=C1 |
| C7H8N2O2 | |
| MFCD00955996 | |
| methyl 4-aminonicotinate, 4-aminonicotinic acid methyl ester, 4-amino-nicotinic acid methyl ester, 4-aminopyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylic acid, 4-amino-, methyl ester, nicotinic acid, 4-amino-, methyl ester, 4-aminonicotinicacidmethylester, methyl-4-aminonicotinate, ksc493a3l, tpc-py097 | |
| methyl 4-aminopyridine-3-carboxylate |
Specifications
| 16135-36-7 | |
| White | |
| MFCD00955996 | |
| methyl 4-aminonicotinate, 4-aminonicotinic acid methyl ester, 4-amino-nicotinic acid methyl ester, 4-aminopyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylic acid, 4-amino-, methyl ester, nicotinic acid, 4-amino-, methyl ester, 4-aminonicotinicacidmethylester, methyl-4-aminonicotinate, ksc493a3l, tpc-py097 | |
| COC(=O)C1=C(N)C=CN=C1 | |
| 152.15 | |
| 152.15 | |
| Crystalline Powder |
| 174°C | |
| C7H8N2O2 | |
| 1 g | |
| OCARFFAPQGYGBP-UHFFFAOYSA-N | |
| methyl 4-aminopyridine-3-carboxylate | |
| 592353 | |
| ≥98.0% (GC,T) | |
| Methyl 4-Aminonicotinate |
Safety and Handling
TSCA : No