missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Aminonicotinate 98.0+%, TCI America™

Catalog No. M23031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23031G 1 g
M23035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23031G Supplier TCI America Supplier No. M23031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16135-36-7 | C7H8N2O2 | 152.15 g/mol

Chemical Identifiers

CAS 16135-36-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00955996
InChI Key OCARFFAPQGYGBP-UHFFFAOYSA-N
Synonym methyl 4-aminonicotinate, 4-aminonicotinic acid methyl ester, 4-amino-nicotinic acid methyl ester, 4-aminopyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylic acid, 4-amino-, methyl ester, nicotinic acid, 4-amino-, methyl ester, 4-aminonicotinicacidmethylester, methyl-4-aminonicotinate, ksc493a3l, tpc-py097
PubChem CID 592353
IUPAC Name methyl 4-aminopyridine-3-carboxylate
SMILES COC(=O)C1=C(N)C=CN=C1

Specifications

Melting Point 174°C
Color White
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Aminonicotinate