missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Aminomethyl)cyclohexanecarboxylate Hydrochloride (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M19871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19871G 1 g
M19875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19871G Supplier TCI America Supplier No. M19871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54640-02-7
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD03093748
InChI Key PZPSRTWFJCGJLP-UHFFFAOYSA-N
Synonym 4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 12721445
IUPAC Name methyl 4-(aminomethyl)cyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)CN.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 207.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Aminomethyl)cyclohexanecarboxylate Hydrochloride (cis- and trans- mixture)