missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Aminocyclohexanecarboxylate Hydrochloride (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M19845G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19845G Supplier TCI America Supplier No. M19845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent for research applications, supplied as a cis- and trans- isomer mixture with ≥98.0% purity.

Chemical Identifiers

CAS 100707-54-8
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.671
InChI Key NHAYDXCUCXRAMF-UHFFFAOYSA-N
Synonym 4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 18521575
IUPAC Name methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)N.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 193.67
Percent Purity ≥98.0% (N,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Aminocyclohexanecarboxylate Hydrochloride (cis- and trans- mixture)