missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,6-dichloronicotinate, 95%

Catalog No. AAH6407706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6407706 5 g
AAH6407714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6407706 Supplier Thermo Scientific Chemicals Supplier No. H6407706
Only null left

CAS: 65973-52-6 | C7H5Cl2NO2 | 206.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65973-52-6
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD04125732
InChI Key JEJMDUMJSZTJTI-UHFFFAOYSA-N
PubChem CID 2764083
IUPAC Name methyl 4,6-dichloropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(Cl)C=C1Cl

Specifications

Quantity 5 g
Beilstein 13279782
Synonym methyl 4,6-dichloronicotinate, methyl4,6-dichloronicotinate, 4,6-dichloro-nicotinic acid methyl, 4,6-dichloro-nicotinic acid methyl ester, 4,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, methyl ester, 2-p-tolylimidazo 1,2-a pyridine, pubchem12979, ksc494c6d, methyl-4,6-dichloronicotinate
Formula Weight 206.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 4,6-dichloronicotinate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only