missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAB2078006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2078006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2078006 Supplier Thermo Scientific Chemicals Supplier No. B2078006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96232-71-2 | Molecular Formula: C6H4Br2O3S | Molecular Weight: 315.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96232-71-2
Molecular Formula C6H4Br2O3S
Molecular Weight (g/mol) 315.96
MDL Number MFCD00052082
InChI Key YZPNGCXFJCPJSV-XQRVVYSFSA-N
Synonym methyl4,5-dibromo-3-hydroxythiophene-2-carboxylate, 4,5-dibromo-3-hydroxythiophene-2-carboxylic acid methyl ester, 2z-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, acmc-20akgp, maybridge1_003706, methyl 4,5-dibromo-3-hydroxy-2-thiophenecarboxylate, 4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylat, 2-thiophenecarboxylicacid,4,5-dibromo-3-hydroxy-,methyl ester, 2e-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one
PubChem CID 2777613
IUPAC Name methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate
SMILES CO\C(O)=C1/SC(Br)=C(Br)C1=O

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Formula Weight 316.89
Percent Purity 97%
Chemical Name or Material Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only