missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,4,4-trifluorocrotonate, 97%

Catalog No. p-7058373
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1146603 1 g
AAL1146606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1146603 Supplier Thermo Scientific Chemicals Supplier No. L1146603

CAS: 85694-31-1 | C5H5F3O2 | 154.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85694-31-1
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.09
MDL Number MFCD00077567
InChI Key DMMZYYLXAGRBDO-NSCUHMNNSA-N
Synonym methyl 4,4,4-trifluorocrotonate, methyl 2e-4,4,4-trifluorobut-2-enoate, methyl4,4,4-trifluorocrotonate, methyl e-3-trifluoromethyl propenoate, e-methyl 4,4,4-trifluorobut-2-enoate, methyl 4,4,4-trifluorobut-2-enoate, heddpndiaicichibemultbb`, methyl 4,4,4-trifluorocrotonat, methyl e-4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid methyl ester
PubChem CID 5371282
IUPAC Name methyl (E)-4,4,4-trifluorobut-2-enoate
SMILES COC(=O)\C=C\C(F)(F)F

Specifications

Boiling Point 94°C to 95°C
Refractive Index 1.3535
Quantity 1 g
UN Number UN3272
Formula Weight 154.1
Percent Purity 97%
Chemical Name or Material Methyl 4,4,4-trifluorocrotonate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only