missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 209912-44-7
Molecular Formula C11H10N2O3
Molecular Weight (g/mol) 218.212
MDL Number MFCD08690288
InChI Key PJYMNVKGDLTDJE-UHFFFAOYSA-N
Synonym methyl 4-3-methyl-1,2,4-oxadiazol-5-yl benzoate, 4-3-methyl-1,2,4-oxadiazole-5-yl benzoic acid methyl ester, benzoicacid, 4-3-methyl-1,2,4-oxadiazol-5-yl-, methyl ester
PubChem CID 11096067
IUPAC Name methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate
SMILES CC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC50922CB
SDS View Documents
Thermo Scientific Chemicals
CC50922CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50922DA
SDS View Documents
Thermo Scientific Chemicals
CC50922DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

209912-44-7
218.212
PJYMNVKGDLTDJE-UHFFFAOYSA-N
11096067
CC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC
C11H10N2O3
MFCD08690288
methyl 4-3-methyl-1,2,4-oxadiazol-5-yl benzoate, 4-3-methyl-1,2,4-oxadiazole-5-yl benzoic acid methyl ester, benzoicacid, 4-3-methyl-1,2,4-oxadiazol-5-yl-, methyl ester
methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate

Specifications

209912-44-7
100.0
Amber Glass Bottle
MFCD08690288
methyl 4-3-methyl-1,2,4-oxadiazol-5-yl benzoate, 4-3-methyl-1,2,4-oxadiazole-5-yl benzoic acid methyl ester, benzoicacid, 4-3-methyl-1,2,4-oxadiazol-5-yl-, methyl ester
CC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC
218.212
218.21
Methyl 4-(3-methyl-1, 2, 4-oxadiazol-5-yl)benzoate
96.5
97%
C11H10N2O3
250mg
PJYMNVKGDLTDJE-UHFFFAOYSA-N
methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate
11096067
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents