missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.

CAS: 79322-76-2 | C10H12O3 | 180.2 g/mol

$693.62 - $693.62

Chemical Identifiers

CAS 79322-76-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00216476
InChI Key KAXLTAULVFFCNL-UHFFFAOYSA-N
PubChem CID 586417
IUPAC Name methyl 4-(1-hydroxyethyl)benzoate
SMILES CC(C1=CC=C(C=C1)C(=O)OC)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424750050
SDS View Documents
Thermo Scientific Chemicals
424750050
5 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

79322-76-2
180.2
KAXLTAULVFFCNL-UHFFFAOYSA-N
methyl 4-(1-hydroxyethyl)benzoate
C10H12O3
MFCD00216476
586417
CC(C1=CC=C(C=C1)C(=O)OC)O

Specifications

79322-76-2
148°C to 152°C (1.0 mmHg)
Authentic
Glass bottle
1.5320 to 1.536
5 g
Solubility in water: slightly
CC(C1=CC=C(C=C1)C(=O)OC)O
180.2
180.2
Technical
Methyl 4-(1-hydroxyethyl)benzoate
Colorless to Yellow
>93°C
88% min. (GC)
C10H12O3
MFCD00216476
methyl 4-1-hydroxyethyl benzoate, 4-1-hydroxyethyl benzoic acid methyl ester, 4-1-hydroxy-ethyl-benzoic acid methyl ester, benzoic acid, 4-1-hydroxyethyl-, methyl ester, acmc-20m5ni, benzoic acid, 4-1s-1-hydroxyethyl-, methyl ester, methyl 4-hydroxyethyl benzoate, methyl4-1-hydroxyethyl benzoate, methyl-4-1-hydroxyethyl benzoate, methyl 4-1-hydroxyethyl benzoate #
KAXLTAULVFFCNL-UHFFFAOYSA-N
methyl 4-(1-hydroxyethyl)benzoate
586417
90%
Viscous Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only