missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.

Catalog No. AC424750050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424750050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424750050 Supplier Thermo Scientific Chemicals Supplier No. 424750050
Only null left

CAS: 79322-76-2 | C10H12O3 | 180.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79322-76-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00216476
InChI Key KAXLTAULVFFCNL-UHFFFAOYSA-N
PubChem CID 586417
IUPAC Name methyl 4-(1-hydroxyethyl)benzoate
SMILES CC(C1=CC=C(C=C1)C(=O)OC)O

Specifications

Color Colorless to Yellow
Boiling Point 148°C to 152°C (1.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.536
Quantity 5 g
Synonym methyl 4-1-hydroxyethyl benzoate, 4-1-hydroxyethyl benzoic acid methyl ester, 4-1-hydroxy-ethyl-benzoic acid methyl ester, benzoic acid, 4-1-hydroxyethyl-, methyl ester, acmc-20m5ni, benzoic acid, 4-1s-1-hydroxyethyl-, methyl ester, methyl 4-hydroxyethyl benzoate, methyl4-1-hydroxyethyl benzoate, methyl-4-1-hydroxyethyl benzoate, methyl 4-1-hydroxyethyl benzoate #
Solubility Information Solubility in water: slightly
Formula Weight 180.2
Percent Purity 90%
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material Methyl 4-(1-hydroxyethyl)benzoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only