missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Phenylpropionate 98.0+%, TCI America™

Catalog No. M178625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M178625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M178625G Supplier TCI America Supplier No. M178625G

Chemical Identifiers

CAS 103-25-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00017209
InChI Key RPUSRLKKXPQSGP-UHFFFAOYSA-N
Synonym methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
PubChem CID 7643
IUPAC Name methyl 3-phenylpropanoate
SMILES COC(=O)CCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 239°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Phenylpropionate
EINECSNumber (3)-1730
TSCA Yes