Learn More
Methyl 3-phenylpropionate, 98%
CAS: 103-25-3 | C10H12O2 | 164.204 g/mol
$180.75 - $674.70
Chemical Identifiers
CAS | 103-25-3 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.204 |
MDL Number | MFCD00017209 |
InChI Key | RPUSRLKKXPQSGP-UHFFFAOYSA-N |
Synonym | methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10Show More |
PubChem CID | 7643 |
IUPAC Name | methyl 3-phenylpropanoate |
SMILES | COC(=O)CCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0531018
|
Thermo Scientific Chemicals
L0531018 |
50 g |
Each for $180.75
|
|
||||||
AAL0531030
|
Thermo Scientific Chemicals
L0531030 |
250 g |
Each for $674.70
|
|
||||||
Description
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
103-25-3 | |
164.204 | |
RPUSRLKKXPQSGP-UHFFFAOYSA-N | |
7643 | |
COC(=O)CCC1=CC=CC=C1 |
C10H12O2 | |
MFCD00017209 | |
methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10Show More | |
methyl 3-phenylpropanoate |
Specifications
103-25-3 | |
236°C to 239°C | |
C10H12O2 | |
MFCD00017209 | |
1909591 | |
Soluble in chloroform. Insoluble in water. | |
COC(=O)CCC1=CC=CC=C1 | |
164.204 | |
164.2 | |
Methyl 3-phenylpropionate |
1.045 | |
100°C (212°F) | |
1.502 | |
50 g | |
methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10 | |
RPUSRLKKXPQSGP-UHFFFAOYSA-N | |
methyl 3-phenylpropanoate | |
7643 | |
98% |
Safety and Handling
EINECSNumber : 203-092-2
TSCA : Yes
RUO – Research Use Only