missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%

Catalog No. p-7055797
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6446706 5 g
AAH6446714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6446706 Supplier Thermo Scientific Chemicals Supplier No. H6446706
Only null left

CAS: 188614-01-9 | C10H9NO3S | 223.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188614-01-9
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00449104
InChI Key UFSVRHNRNVVBJR-UHFFFAOYSA-N
PubChem CID 699501
IUPAC Name methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
SMILES COC(=O)C1=CC2=C(SCC(=O)N2)C=C1

Specifications

Melting Point 97°C to 99°C
Quantity 5 g
Beilstein 14311523
Synonym methyl 3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate, methyl-3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate, methyl 3-oxo-3,4-dihydro-2h-benzo b 1,4 thiazine-6-carboxylate, methyl 3-oxo-2h-1,4-benzothiazine-6-carboxylate, methyl 3,4-dihydro-3-oxo-2h-benzo b 1,4 thiazine-6-carboxylate, methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate, pubchem13044, ksc497g3b, methyl-3-oxo-3,4-dihydro-2h-1,, methyl3-oxo-2h-1,4-benzothiazine-6-carboxylate
Formula Weight 223.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only