missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrosalicylate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 22621-41-6
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00272263
InChI Key NIBVYEHAFBEVFI-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
PubChem CID 596904
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC381920050
View Documents
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

22621-41-6
197.15
NIBVYEHAFBEVFI-UHFFFAOYSA-N
596904
C8H7NO5
MFCD00272263
methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O

Specifications

22621-41-6
97.5% min. (GC)
NO2C6H3(OH)COOCH3
methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
596904
98%
Authentic
C8H7NO5
MFCD00272263
NIBVYEHAFBEVFI-UHFFFAOYSA-N
197.15
197.15
Methyl 3-nitrosalicylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only