missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Nitrophenylacetate 98.0+%, TCI America™

Catalog No. M25221G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25221G Supplier TCI America Supplier No. M25221G
Only null left

Chemical Identifiers

CAS 10268-12-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD08669939
InChI Key BFGYITRIATVARH-UHFFFAOYSA-N
Synonym 3-Nitrophenylacetic Acid Methyl Ester
PubChem CID 526967
IUPAC Name methyl 2-(3-nitrophenyl)acetate
SMILES COC(=O)CC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 28°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Nitrophenylacetate