missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methylthiophene-2-carboxylate, 99%

Catalog No. p-7047225
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAL1809303 1 g
AAL1809306 5 g
2 options

Catalog No. AAL1809303

Supplier: Thermo Scientific Chemicals L1809303

Only null left

CAS: 81452-54-2 | C7H8O2S | 156.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81452-54-2
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD00234332
InChI Key BRWROFVPMUPMJQ-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-thiophenecarboxylate, 3-methylthiophene-2-carboxylic acid methyl ester, 3-methyl-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-methyl-, methyl ester, methyl 3-methyl-thiophene-2-carboxylate, d methyl ester, 3-methyl-thiophene-2-carboxylic acid, pubchem10098, pubchem22915, ksc495k9b
PubChem CID 580757
IUPAC Name methyl 3-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C)C=CS1

Specifications

Boiling Point 216°C to 217°C
Refractive Index 1.5168
Quantity 1 g
Beilstein 116836
Formula Weight 156.2
Percent Purity 99%
Chemical Name or Material Methyl 3-methylthiophene-2-carboxylate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only