missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methylphenylacetate, 98%

Catalog No. p-3212874
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC374080050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC374080050 Supplier Thermo Scientific Chemicals Supplier No. 374080050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53088-69-0 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53088-69-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00968468
InChI Key AWTKFTNNPQGGLX-UHFFFAOYSA-N
Synonym methyl 2-m-tolyl acetate, methyl 3-methylphenylacetate, methyl 2-3-methylphenyl acetate, 3-methylphenylacetic acid methyl ester, benzeneacetic acid, 3-methyl-, methyl ester, methyl 2-m-tolylacetate, m-tolylacetic acid methyl ester, m-toly-acetic acid methyl ester, m-tolyl-acetic acid methyl ester, methyl 2-3-methylphenyl ethanoate
PubChem CID 5135288
IUPAC Name methyl 2-(3-methylphenyl)acetate
SMILES COC(=O)CC1=CC=CC(C)=C1

Specifications

Color Colorless to Yellow
Boiling Point 117°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5045 to 1.5065
Linear Formula CH3C6H4CH2COOCH3
Quantity 5 g
Formula Weight 164.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-methylphenylacetate
Show More Show Less

RUO – Research Use Only