missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Mercaptopropionate 98.0+%, TCI America™

Catalog No. M143425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M143425G 25 g
M1434500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M143425G Supplier TCI America Supplier No. M143425G

Chemical Identifiers

CAS 2935-90-2
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00004895
InChI Key LDTLDBDUBGAEDT-UHFFFAOYSA-N
Synonym methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
PubChem CID 18050
IUPAC Name methyl 3-sulfanylpropanoate
SMILES COC(=O)CCS

Specifications

Color Yellow
Quantity 25 g
UN Number 2810
Formula Weight 120.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Mercaptopropionate
EINECSNumber (2)-3062
RTECSNumber UA2465980
TSCA Yes