missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-mercaptopropionate, 98%

Catalog No. AAB2071822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2071822 100 g
AAB2071814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2071822 Supplier Thermo Scientific Chemicals Supplier No. B2071822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 2935-90-2 | C4H8O2S | 120.166 g/mol

Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2935-90-2
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00004895
InChI Key LDTLDBDUBGAEDT-UHFFFAOYSA-N
Synonym methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
PubChem CID 18050
IUPAC Name methyl 3-sulfanylpropanoate
SMILES COC(=O)CCS

Specifications

Density 1.085
Boiling Point 164°C
Flash Point 60°C (140°F)
Refractive Index 1.463
Quantity 100 g
UN Number UN1228
Beilstein 1742952
Solubility Information Soluble in water.
Formula Weight 120.17
Percent Purity 98%
Chemical Name or Material Methyl 3-mercaptopropionate
Show More Show Less
Hazard Category H226-H301-H312-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H331-H226-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 220-912-4
RTECSNumber UA2465980
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only