missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxyisoxazole-5-carboxylate 98.0+%, TCI America™

Catalog No. p-7133223
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24411G 1 g
M24415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24411G Supplier TCI America Supplier No. M24411G
Only null left

Chemical Identifiers

CAS 10068-07-2
Molecular Formula C5H5NO4
Molecular Weight (g/mol) 143.10
MDL Number MFCD00062870
InChI Key BBFWUUBQSXVHHZ-UHFFFAOYSA-N
Synonym methyl 3-hydroxyisoxazole-5-carboxylate, methyl 3-hydroxy-5-isoxazolecarboxylate, methyl 3-hydroxy-1,2-oxazole-5-carboxylate, 3-hydroxyisoxazole-5-carboxylic acid methyl ester, methyl 3-hydroxyl-5-isoxazolecarboxylate, methyl-3-hydroxy-5-isoxazolecarboxylate, 5-isoxazolecarboxylic acid, 2,3-dihydro-3-oxo-, methyl ester, 3-hydroxy-isoxazole-5-carboxylic acid methyl ester, methyl 3-oxo-2,3-dihydro-1,2-oxazole-5-carboxylate, methyl 3-oxo-2h-1,2-oxazole-5-carboxylate
PubChem CID 2724585
IUPAC Name methyl 3-oxo-2,3-dihydro-1,2-oxazole-5-carboxylate
SMILES COC(=O)C1=CC(=O)NO1

Specifications

Melting Point 164°C
Color Yellow
Quantity 1 g
Formula Weight 143.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Hydroxyisoxazole-5-carboxylate