missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-4-methoxybenzoate, 98%, Thermo Scientific™

Catalog No. AC399260050
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399260050 5g
AC399260010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399260050 Supplier Thermo Scientific Chemicals Supplier No. 399260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6702-50-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD01321262
InChI Key QXOXUEFXRSIYSW-UHFFFAOYSA-N
Synonym methyl isovanillate, methyl 3-hydroxy-4-methoxybenzenecarboxylate, 3-hydroxy-4-methoxybenzoic acid methyl ester, isovanillic acid methyl ester, methyl-3-hydroxy-4-methoxybenzoate, methyl 3-hydroxy-4-methoxy-benzoate, benzoic acid, 3-hydroxy-4-methoxy-, methyl ester, methyl 3-hydroxy-4-methoxy benzoate, isovamllic acid methyl ester, ksc496e5l
PubChem CID 4056967
IUPAC Name methyl 3-hydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC(O)=C(OC)C=C1

Specifications

Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)COOCH3
Quantity 5g
Formula Weight 182.17
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxy-4-methoxybenzoate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only