missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Hydroxy-2-naphthoate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 75°C |
| Color | Yellow |
| Boiling Point | 207°C |
| Quantity | 25 g |
| Formula Weight | 202.21 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 3-Hydroxy-2-naphthoate |
Safety and Handling
| EINECSNumber | (4)-0400 |
| TSCA | Yes |