missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hexenoate, 95+%, predominantly trans, Thermo Scientific™

Catalog No. p-3593457
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393550050 5g Glass bottle
AC393550250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393550050 Supplier Thermo Scientific Chemicals Supplier No. 393550050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2396-78-3
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00672771
InChI Key XEAIHUDTEINXFG-SNAWJCMRSA-N
Synonym 3-hexenoic acid, methyl ester, methyl 3-hexenoate, methyl trans-3-hexenoate, methyl e-hex-3-enoate, honeyflor, 3-hexenoic acid, methyl ester, e, unii-7ip0t834pb, methyl hex-3-enoate, methyl e-3-hexenoate, methyl trans 3 hexenoate
PubChem CID 5362782
IUPAC Name methyl (E)-hex-3-enoate
SMILES CCC=CCC(=O)OC

Specifications

Density 0.9150g/mL
Boiling Point 169°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Linear Formula CH3CH2CHCHCH2COOCH3
Quantity 5g
Specific Gravity 0.915
Solubility Information Solubility in water: practically insoluble
Formula Weight 128.17
Percent Purity 95+%
Chemical Name or Material Methyl 3-hexenoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-256-1

RUO – Research Use Only