missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-fluorophenylacetate, 98%, Thermo Scientific™

Catalog No. p-163287
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC351090050 5g Glass bottle
AC351090250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC351090050 Supplier Thermo Scientific Chemicals Supplier No. 351090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 64123-77-9
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD03093288
InChI Key OXYLCGRXCQARQV-UHFFFAOYSA-N
Synonym methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
PubChem CID 2734108
IUPAC Name methyl 2-(3-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=CC(F)=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.489
Linear Formula FC6H4CH2CO2H
Quantity 5g
Formula Weight 168.17
Percent Purity 98%
Chemical Name or Material Methyl 3-fluorophenylacetate, 98%

RUO – Research Use Only