missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(Dimethylamino)benzoate 99.0+%, TCI America™
$164.03 - $326.21
Chemical Identifiers
| CAS | 16518-64-2 |
|---|---|
| Molecular Formula | C10H13NO2 |
| Molecular Weight (g/mol) | 179.22 |
| MDL Number | MFCD00144731 |
| InChI Key | CABFTHPIDKWPNQ-UHFFFAOYSA-N |
| Synonym | 3-(Dimethylamino)benzoic Acid Methyl Ester |
| PubChem CID | 3627900 |
| IUPAC Name | methyl 3-(dimethylamino)benzoate |
| SMILES | COC(=O)C1=CC=CC(=C1)N(C)C |
Chemical Identifiers
| 16518-64-2 | |
| 179.22 | |
| CABFTHPIDKWPNQ-UHFFFAOYSA-N | |
| 3627900 | |
| COC(=O)C1=CC=CC(=C1)N(C)C |
| C10H13NO2 | |
| MFCD00144731 | |
| 3-(Dimethylamino)benzoic Acid Methyl Ester | |
| methyl 3-(dimethylamino)benzoate |
Specifications
| 16518-64-2 | |
| 80°C | |
| MFCD00144731 | |
| 3-(Dimethylamino)benzoic Acid Methyl Ester | |
| COC(=O)C1=CC=CC(=C1)N(C)C | |
| 179.22 | |
| 179.22 | |
| Liquid |
| Yellow | |
| C10H13NO2 | |
| 10 g | |
| CABFTHPIDKWPNQ-UHFFFAOYSA-N | |
| methyl 3-(dimethylamino)benzoate | |
| 3627900 | |
| ≥99.0% (T) | |
| Methyl 3-(Dimethylamino)benzoate |
Safety and Handling
EINECSNumber : (3)-2999
TSCA : No