missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(Dimethylamino)benzoate 99.0+%, TCI America™

Catalog No. D206910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D206910G 10 g
D206925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D206910G Supplier TCI America Supplier No. D206910G
Only null left

Chemical Identifiers

CAS 16518-64-2
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00144731
InChI Key CABFTHPIDKWPNQ-UHFFFAOYSA-N
Synonym 3-(Dimethylamino)benzoic Acid Methyl Ester
PubChem CID 3627900
IUPAC Name methyl 3-(dimethylamino)benzoate
SMILES COC(=O)C1=CC=CC(=C1)N(C)C

Specifications

Color Yellow
Boiling Point 80°C
Quantity 10 g
Formula Weight 179.22
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl 3-(Dimethylamino)benzoate
EINECSNumber (3)-2999
TSCA No