missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(chlorosulfonyl)-4-methoxybenzoate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 192323-12-9
Molecular Formula C9H9ClO5S
Molecular Weight (g/mol) 264.68
MDL Number MFCD03407355
InChI Key FKAIQWMEWVOGDP-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl-4-methoxybenzoate, methyl 3-chlorosulfonyl-4methoxybenzoate, 2-chlorosulphonyl-4-methoxycarbonyl anisole, methyl 3-chlorosulfonyl-4-methyloxy benzoate, methyl-3-chlorosulfonyl-4-methoxybenzoate, 2-methoxy-5-methoxycarbonyl benzenesulphonyl chloride, benzoic acid,3-chlorosulfonyl-4-methoxy-, methyl ester
PubChem CID 2794613
IUPAC Name methyl 3-chlorosulfonyl-4-methoxybenzoate
SMILES COC(=O)C1=CC(=C(OC)C=C1)S(Cl)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MO00348DA
SDS View Documents
Thermo Scientific Chemicals
MO00348DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

192323-12-9
264.68
FKAIQWMEWVOGDP-UHFFFAOYSA-N
2794613
COC(=O)C1=CC(=C(OC)C=C1)S(Cl)(=O)=O
C9H9ClO5S
MFCD03407355
methyl 3-chlorosulfonyl-4-methoxybenzoate, methyl 3-chlorosulfonyl-4methoxybenzoate, 2-chlorosulphonyl-4-methoxycarbonyl anisole, methyl 3-chlorosulfonyl-4-methyloxy benzoate, methyl-3-chlorosulfonyl-4-methoxybenzoate, 2-methoxy-5-methoxycarbonyl benzenesulphonyl chloride, benzoic acid,3-chlorosulfonyl-4-methoxy-, methyl ester
methyl 3-chlorosulfonyl-4-methoxybenzoate

Specifications

192323-12-9
97%
C9H9ClO5S
MFCD03407355
methyl 3-chlorosulfonyl-4-methoxybenzoate, methyl 3-chlorosulfonyl-4methoxybenzoate, 2-chlorosulphonyl-4-methoxycarbonyl anisole, methyl 3-chlorosulfonyl-4-methyloxy benzoate, methyl-3-chlorosulfonyl-4-methoxybenzoate, 2-methoxy-5-methoxycarbonyl benzenesulphonyl chloride, benzoic acid,3-chlorosulfonyl-4-methoxy-, methyl ester
COC(=O)C1=CC(=C(OC)C=C1)S(Cl)(=O)=O
264.68
264.68
Methyl 3-(chlorosulfonyl)-4-methoxybenzoate
97.0
Amber glass bottle
CH3OC6H3(C(O)OCH3)S(Cl)O2
1g
FKAIQWMEWVOGDP-UHFFFAOYSA-N
methyl 3-chlorosulfonyl-4-methoxybenzoate
2794613
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'