missing translation for 'onlineSavingsMsg'
Learn More

Methyl (3-Chlorophenyl)acetate 98.0+%, TCI America™

Catalog No. M21425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21425G 5 g
M214225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21425G Supplier TCI America Supplier No. M21425G
Only null left

Chemical Identifiers

CAS 53088-68-9
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00800679
InChI Key HTBBVKFFDDQVER-UHFFFAOYSA-N
PubChem CID 2733544
IUPAC Name methyl 2-(3-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=CC(Cl)=C1

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Synonym methyl 2-3-chlorophenyl acetate, methyl 3-chlorophenylacetate, methyl 3-chlorophenyl acetate, phenylacetic acid, 3-chloro, methyl ester, benzeneacetic acid, 3-chloro-, methyl ester, methyl2-3-chlorophenyl acetate, methyl 2-3-chlorophenyl ethanoate, 3-chlorophenylacetic acid methyl ester, 3-chlorophenyl acetic acid methyl ester
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl (3-Chlorophenyl)acetate